accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNH2

CAS RN 702-82-9

IUPAC Name: 3-aminoadamantan-1-ol
Synonyms: 3-aminoadamantan-1-ol
Molecular Weight (g/mol): 167.25
Molecular Formula: C10H17NO
InChi Key: DWPIPTNBOVJYAD-UHFFFAOYNA-N
SMILES: NC12CC3CC(C1)CC(O)(C3)C2
Molecular Weight (g/mol) 
  • (3)
  • (5)
Percent Purity 
  • (3)
  • (2)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (4)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (3)
  • (5)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (4)
  • (3)