accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOBr

CAS RN 70201-43-3

IUPAC Name: 3-bromopyridine-4-carbaldehyde
Synonyms: 3-bromopyridine-4-carbaldehyde
Molecular Weight (g/mol): 186.01
Molecular Formula: C6H4BrNO
InChi Key: NOBDKWLIAQKADB-UHFFFAOYSA-N
SMILES: BrC1=C(C=O)C=CN=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results