accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 70260-05-8

IUPAC Name: (4-bromothiophen-3-yl)methanol
Synonyms: (4-bromothiophen-3-yl)methanol
Molecular Weight (g/mol): 193.06
Molecular Formula: C5H5BrOS
InChi Key: MPKBPMYJCDNVEF-UHFFFAOYSA-N
SMILES: OCC1=CSC=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results