accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NSN

CAS RN 70291-62-2

IUPAC Name: 2-amino-4H,5H,6H-cyclopenta[b]thiophene-3-carbonitrile
Synonyms: 2-amino-4H,5H,6H-cyclopenta[b]thiophene-3-carbonitrile
Molecular Weight (g/mol): 164.23
Molecular Formula: C8H8N2S
InChi Key: RILAXFOADRDGPQ-UHFFFAOYSA-N
SMILES: NC1=C(C#N)C2=C(CCC2)S1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)