accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHOCH3O

CAS RN 703-23-1

IUPAC Name: 1-(2-hydroxy-6-methoxyphenyl)ethan-1-one
Synonyms: 1-(2-hydroxy-6-methoxyphenyl)ethanone
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: UENLHUMCIOWYQN-UHFFFAOYSA-N
SMILES: COC1=CC=CC(O)=C1C(C)=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results