accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClHNO

CAS RN 703-82-2

IUPAC Name: 6-chloro-1H-indole-3-carbaldehyde
Synonyms: 6-chloro-1H-indole-3-carbaldehyde
Molecular Weight (g/mol): 179.6
Molecular Formula: C9H6ClNO
InChi Key: CTNIXLBHXMSZKL-UHFFFAOYSA-N
SMILES: ClC1=CC=C2C(NC=C2C=O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results