accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7031-23-4

IUPAC Name: 3-(methylsulfanyl)propanoyl chloride
Synonyms: 3-(methylsulfanyl)propanoyl chloride
Molecular Weight (g/mol): 138.61
Molecular Formula: C4H7ClOS
InChi Key: NRHMXMBVLXSSAX-UHFFFAOYSA-N
SMILES: CSCCC(Cl)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results