accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNNNOOI

CAS RN 70315-70-7

IUPAC Name: 3-iodo-6-nitro-2H-indazole
Synonyms: 3-iodo-6-nitro-2H-indazole
Molecular Weight (g/mol): 289.03
Molecular Formula: C7H4IN3O2
InChi Key: GZCGNGLOCQEDMT-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC2=NNC(I)=C2C=C1
Quantity 
  • (1)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results