accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 70326-37-3

IUPAC Name: 3-benzoyl-1,3-thiazolidine-2-thione
Synonyms: 3-benzoyl-1,3-thiazolidine-2-thione
Molecular Weight (g/mol): 223.31
Molecular Formula: C10H9NOS2
InChi Key: NWKQMWLTQNBOHI-UHFFFAOYSA-N
SMILES: O=C(N1CCSC1=S)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results