CAS RN 7035-09-8
IUPAC Name:
5-chloro-2-methoxybenzaldehyde
Synonyms:
5-chloro-2-methoxybenzaldehyde
Molecular Weight (g/mol):
170.59
Molecular Formula:
C8H7ClO2
InChi Key:
HXTWDGGMXZXOIV-UHFFFAOYSA-N
SMILES:
COC1=C(C=O)C=C(Cl)C=C1
1
–
1
of
1
results
1