accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 70351-86-9

IUPAC Name: 2,3,6,7,10,11-hexakis(hexyloxy)triphenylene
Synonyms: 2,3,6,7,10,11-hexakis(hexyloxy)triphenylene
Molecular Weight (g/mol): 829.26
Molecular Formula: C54H84O6
InChi Key: SQYCPYUKCAKHRN-UHFFFAOYSA-N
SMILES: CCCCCCOC1=C(OCCCCCC)C=C2C(=C1)C1=CC(OCCCCCC)=C(OCCCCCC)C=C1C1=CC(OCCCCCC)=C(OCCCCCC)C=C21
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results