accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNO

CAS RN 70380-73-3

IUPAC Name: 2-(1,3-oxazol-5-yl)pyridine
Synonyms: 2-(1,3-oxazol-5-yl)pyridine
Molecular Weight (g/mol): 146.15
Molecular Formula: C8H6N2O
InChi Key: LUMKXMDGONYZHO-UHFFFAOYSA-N
SMILES: O1C=NC=C1C1=CC=CC=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results