accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CONONHHOSOO

CAS RN 704888-90-4

IUPAC Name: N-hydroxy-2-[N-(propan-2-yloxy)[1,1'-biphenyl]-4-sulfonamido]acetamide
Synonyms: N-hydroxy-2-{N-isopropoxy[1,1'-biphenyl]-4-sulfonamido}acetamide
Molecular Weight (g/mol): 364.42
Molecular Formula: C17H20N2O5S
InChi Key: PHGLPDURIUEELR-UHFFFAOYSA-N
SMILES: CC(C)ON(CC(=O)NO)S(=O)(=O)C1=CC=C(C=C1)C1=CC=CC=C1
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results