accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 704890-84-6

IUPAC Name: 2-({5'-bromo-[2,2'-bithiophen]-5-yl}methylidene)propanedinitrile
Synonyms: 2-({5'-bromo-[2,2'-bithiophen]-5-yl}methylidene)propanedinitrile
Molecular Weight (g/mol): 321.21
Molecular Formula: C12H5BrN2S2
InChi Key: GZLRCTASJSGPHL-UHFFFAOYSA-N
SMILES: BrC1=CC=C(S1)C1=CC=C(S1)C=C(C#N)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results