accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClSOOCl

CAS RN 705-21-5

IUPAC Name: 3,5-dichlorobenzene-1-sulfonyl chloride
Synonyms: 3,5-dichlorobenzenesulfonyl chloride
Molecular Weight (g/mol): 245.5
Molecular Formula: C6H3Cl3O2S
InChi Key: RJSQINMKOSOUGT-UHFFFAOYSA-N
SMILES: ClC1=CC(=CC(Cl)=C1)S(Cl)(=O)=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)