accessibility menu, dialog, popup

UserName

Viewing profile as user:
CHFFF

CAS RN 705-31-7

IUPAC Name: 1-ethynyl-4-(trifluoromethyl)benzene
Synonyms: 1-ethynyl-4-(trifluoromethyl)benzene
Molecular Weight (g/mol): 170.13
Molecular Formula: C9H5F3
InChi Key: XTKBMZQCDBHHKY-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(C=C1)C#C
Boiling Point 
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)