accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 705-60-2

IUPAC Name: [(1E)-2-nitroprop-1-en-1-yl]benzene
Synonyms: phenyl-2-nitropropene
Molecular Weight (g/mol): 163.18
Molecular Formula: C9H9NO2
InChi Key: WGSVFWFSJDAYBM-BQYQJAHWSA-N
SMILES: C\C(=C/C1=CC=CC=C1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)