accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 70555-46-3

IUPAC Name: 6-methoxy-1H-indole-3-carbaldehyde
Synonyms: 6-methoxy-1H-indole-3-carbaldehyde
Molecular Weight (g/mol): 175.19
Molecular Formula: C10H9NO2
InChi Key: JTEFJNIWWXTBMP-UHFFFAOYSA-N
SMILES: COC1=CC=C2C(NC=C2C=O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results