accessibility menu, dialog, popup

UserName

Viewing profile as user:
OH(S)(R)OH(R)ONOHNO

CAS RN 7057-27-4

IUPAC Name: 1-[(2R,3R,5S)-3-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: 1-[(2R,3R,5S)-3-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-pyrimidine-2,4-dione
Molecular Weight (g/mol): 228.2
Molecular Formula: C9H12N2O5
InChi Key: QOXJRLADYHZRGC-SHYZEUOFSA-N
SMILES: OC[C@@H]1C[C@@H](O)[C@@H](O1)N1C=CC(=O)NC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results