CAS RN 7059-70-3
IUPAC Name:
2-[3,5-bis(naphthalen-2-yl)phenyl]naphthalene
Synonyms:
2-[3,5-bis(naphthalen-2-yl)phenyl]naphthalene
Molecular Weight (g/mol):
456.59
Molecular Formula:
C36H24
InChi Key:
AWIOMAWDKDFGDP-UHFFFAOYSA-N
SMILES:
C1=CC2=CC=C(C=C2C=C1)C1=CC(=CC(=C1)C1=CC2=CC=CC=C2C=C1)C1=CC=C2C=CC=CC2=C1
1
–
1
of
1
results
1
1,3,5-Tri(1-naphthyl)benzene 98.0+%, TCI America™
CAS: 7059-70-3 Molecular Formula: C36H24 Molecular Weight (g/mol): 456.588 InChI Key: ZVUZLHDATWCFQW-UHFFFAOYSA-N PubChem CID: 15208120 IUPAC Name: 1-(3,5-dinaphthalen-1-ylphenyl)naphthalene SMILES: C1=CC=C2C(=C1)C=CC=C2C3=CC(=CC(=C3)C4=CC=CC5=CC=CC=C54)C6=CC=CC7=CC=CC=C76