accessibility menu, dialog, popup

UserName

Viewing profile as user:
NN

CAS RN 70591-20-7

IUPAC Name: 2-[(diphenylmethylidene)amino]acetonitrile
Synonyms: 2-[(diphenylmethylidene)amino]acetonitrile
Molecular Weight (g/mol): 220.28
Molecular Formula: C15H12N2
InChi Key: VRLJFRODHVSTIK-UHFFFAOYSA-N
SMILES: N#CCN=C(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)