accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONF(E/Z)

CAS RN 70627-52-0

IUPAC Name: 1-[4-(benzyloxy)phenyl]-N-(4-fluorophenyl)methanimine
Synonyms: 1-[4-(benzyloxy)phenyl]-N-(4-fluorophenyl)methanimine
Molecular Weight (g/mol): 305.35
Molecular Formula: C20H16FNO
InChi Key: IWNBEFDVKWCBFY-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)N=CC1=CC=C(OCC2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results