CAS RN 7064-31-5
IUPAC Name:
5-{[(5Z)-3-(1,5-dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)-4-oxo-2-sulfanylidene-1,3-thiazolidin-5-ylidene]methyl}-4-methyl-6-(3-methylpiperidin-1-yl)-2-oxo-1-propyl-1,2-dihydropyridine-3-carbonitrile
Synonyms:
5-{[(5Z)-3-(1,5-dimethyl-3-oxo-2-phenylpyrazol-4-yl)-4-oxo-2-sulfanylidene-1,3-thiazolidin-5-ylidene]methyl}-4-methyl-6-(3-methylpiperidin-1-yl)-2-oxo-1-propylpyridine-3-carbonitrile
Molecular Weight (g/mol):
602.77
Molecular Formula:
C31H34N6O3S2
InChi Key:
LQPROPLOIBHLEY-XYGWBWBKNA-N
SMILES:
CCCN1C(=O)C(C#N)=C(C)C(\C=C2/SC(=S)N(C2=O)C2=C(C)N(C)N(C2=O)C2=CC=CC=C2)=C1N1CCCC(C)C1
1
–
2
of
2
results
1
Promotions Available
5-(4-Bromophenyl)isoxazole, 98%, Thermo Scientific Chemicals
5-(4-Bromophenyl)isoxazole, 98%, Thermo Scientific Chemicals
CAS: 7064-31-5 Molecular Formula: C9H6BrNO Molecular Weight (g/mol): 224.057 MDL Number: MFCD02183532 InChI Key: AVMMSHZGFRWLRI-UHFFFAOYSA-N Synonym: 5-4-bromophenyl isoxazole,5-4-bromophenyl-1,2-oxazole,acmc-209ofh PubChem CID: 2735608 IUPAC Name: 5-(4-bromophenyl)-1,2-oxazole SMILES: C1=CC(=CC=C1C2=CC=NO2)Br
2
5-(4-Bromophenyl)isoxazole, 97%, Thermo Scientific™
CAS: 7064-31-5 Molecular Formula: C9H6BrNO Molecular Weight (g/mol): 224.057 MDL Number: MFCD02183532 InChI Key: AVMMSHZGFRWLRI-UHFFFAOYSA-N Synonym: 5-4-bromophenyl isoxazole,5-4-bromophenyl-1,2-oxazole,acmc-209ofh PubChem CID: 2735608 IUPAC Name: 5-(4-bromophenyl)-1,2-oxazole SMILES: C1=CC(=CC=C1C2=CC=NO2)Br