CAS RN 707-37-9
IUPAC Name:
3,5-dimethyladamantan-1-ol
Synonyms:
3,5-dimethyladamantan-1-ol
Molecular Weight (g/mol):
180.29
Molecular Formula:
C12H20O
InChi Key:
LBWCITVBZLTEKW-UHFFFAOYNA-N
SMILES:
CC12CC3CC(C)(C1)CC(O)(C3)C2
1
–
1
of
1
results
1
3,5-Dimethyl-1-adamantanol 97.0+%, TCI America™
CAS: 707-37-9 Molecular Formula: C12H20O Molecular Weight (g/mol): 180.291 MDL Number: MFCD00074775 InChI Key: LBWCITVBZLTEKW-UHFFFAOYSA-N Synonym: 1-Hydroxy-3,5-dimethyladamantane PubChem CID: 265793 IUPAC Name: 3,5-dimethyladamantan-1-ol SMILES: CC12CC3CC(C1)(CC(C3)(C2)O)C