CAS RN 707-72-2
IUPAC Name:
1-(dichloromethyl)-2-(trifluoromethyl)benzene
Synonyms:
1-(dichloromethyl)-2-(trifluoromethyl)benzene
Molecular Weight (g/mol):
229.02
Molecular Formula:
C8H5Cl2F3
InChi Key:
JIJFXGFHPXLJME-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=CC=CC=C1C(Cl)Cl
1
–
1
of
1
results
1
2-(Trifluoromethyl)benzal Chloride, TCI America™
CAS: 707-72-2 Molecular Formula: C8H5Cl2F3 Molecular Weight (g/mol): 229.02 MDL Number: MFCD00077613 InChI Key: JIJFXGFHPXLJME-UHFFFAOYSA-N PubChem CID: 606399 IUPAC Name: 1-(dichloromethyl)-2-(trifluoromethyl)benzene SMILES: FC(F)(F)C1=CC=CC=C1C(Cl)Cl