CAS RN 709-50-2
IUPAC Name:
2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
Synonyms:
methyl-α-D-mannoside methyl mannoside mannoside, methyl
Molecular Weight (g/mol):
194.18
Molecular Formula:
C7H14O6
InChi Key:
HOVAGTYPODGVJG-UHFFFAOYNA-N
SMILES:
COC1OC(CO)C(O)C(O)C1O
1
–
1
of
1
results
1
Methyl beta-D-Glucopyranoside Hemihydrate 98.0+%, TCI America™
CAS: 709-50-2 Molecular Formula: C7H14O6 Molecular Weight (g/mol): 194.18 MDL Number: MFCD00006602 InChI Key: HOVAGTYPODGVJG-UHFFFAOYNA-N Synonym: Methyl beta-D-Glucoside PubChem CID: 445238 ChEBI: CHEBI:320055 IUPAC Name: 2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol SMILES: COC1OC(CO)C(O)C(O)C1O