accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 70916-89-1

CH3OOO

CAS RN 70916-89-1

IUPAC Name: methyl 4'-formyl-[1,1'-biphenyl]-4-carboxylate
Synonyms: methyl 4'-formyl-[1,1'-biphenyl]-4-carboxylate
Molecular Weight (g/mol): 240.26
Molecular Formula: C15H12O3
InChi Key: URINKTWOCVLGGR-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(C=C1)C1=CC=C(C=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results