accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 70918-74-0

IUPAC Name: 1-(2,3-dihydro-1,4-benzodioxine-2-carbonyl)piperazine hydrochloride
Synonyms: 1-(2,3-dihydro-1,4-benzodioxine-2-carbonyl)piperazine hydrochloride
Molecular Weight (g/mol): 284.74
Molecular Formula: C13H17ClN2O3
InChi Key: TUKBWYXLYINULI-UHFFFAOYNA-N
SMILES: Cl.O=C(C1COC2=CC=CC=C2O1)N1CCNCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results