accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 70924-78-6

IUPAC Name: N-(2-acetamidocyclohexyl)acetamide
Synonyms: N-(2-acetamidocyclohexyl)acetamide
Molecular Weight (g/mol): 198.27
Molecular Formula: C10H18N2O2
InChi Key: MZYLAHHTXWRMOL-UHFFFAOYNA-N
SMILES: CC(=O)NC1CCCCC1NC(C)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results