accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 709652-82-4

NBrNH2N

CAS RN 709652-82-4

IUPAC Name: 2-amino-5-bromopyridine-3-carbonitrile
Synonyms: 2-amino-5-bromopyridine-3-carbonitrile
Molecular Weight (g/mol): 198.02
Molecular Formula: C6H4BrN3
InChi Key: UJKZMLZIIIGCMI-UHFFFAOYSA-N
SMILES: NC1=NC=C(Br)C=C1C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results