accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3

CAS RN 7098-22-8

IUPAC Name: tetratetracontane
Synonyms: tetratetracontane
Molecular Weight (g/mol): 619.2
Molecular Formula: C44H90
InChi Key: KMXFZRSJMDYPPG-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (1)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)