accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 71-43-2

IUPAC Name: benzene
Synonyms: benzene caswell no. 077 aromatic alkane benzolum benzine (obs.) benzolene BNZ RNG carbon oil benzene (substance)
Molecular Weight (g/mol): 78.11
Molecular Formula: C6H6
InChi Key: UHOVQNZJYSORNB-UHFFFAOYSA-N
SMILES: C1=CC=CC=C1
Concentration 
  • (1)
Element 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)