accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 71-44-3

IUPAC Name: (3-aminopropyl)({4-[(3-aminopropyl)amino]butyl})amine
Synonyms: spermine spermine dihydrate terpene hydrocarbons
Molecular Weight (g/mol): 202.35
Molecular Formula: C10H26N4
InChi Key: PFNFFQXMRSDOHW-UHFFFAOYSA-N
SMILES: NCCCNCCCCNCCCN
Percent Purity 
  • (2)
  • (1)
  • (4)
Quantity 
  • (3)
  • (3)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (4)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)