accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 71022-43-0

IUPAC Name: (3,5-dinitrophenyl)methanol
Synonyms: (3,5-dinitrophenyl)methanol
Molecular Weight (g/mol): 198.13
Molecular Formula: C7H6N2O5
InChi Key: GPHYIQCSMDYRGJ-UHFFFAOYSA-N
SMILES: OCC1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)