accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 71026-66-9

IUPAC Name: tert-butyl N-(4-aminophenyl)carbamate
Synonyms: tert-butyl N-(4-aminophenyl)carbamate
Molecular Weight (g/mol): 208.26
Molecular Formula: C11H16N2O2
InChi Key: WIVYTYZCVWHWSH-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1=CC=C(N)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)