accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH2MgBr

CAS RN 7103-09-5

IUPAC Name: bromo(but-3-en-1-yl)magnesium
Synonyms: bromo(but-3-en-1-yl)magnesium
Molecular Weight (g/mol): 159.31
Molecular Formula: C4H7BrMg
InChi Key: SMLLHQBZPPSOSR-UHFFFAOYSA-M
SMILES: Br[Mg]CCC=C
Molecular Weight (g/mol) 
  • (1)
  • (2)
  • (1)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)