CAS RN 710348-69-9
IUPAC Name:
5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydro-1H-1,3-benzodiazol-2-one
Synonyms:
5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-dihydro-1,3-benzodiazol-2-one
Molecular Weight (g/mol):
260.1
Molecular Formula:
C13H17BN2O3
InChi Key:
LQVSXEXWXILGBH-UHFFFAOYSA-N
SMILES:
CC1(C)OB(OC1(C)C)C1=CC=C2NC(=O)NC2=C1
1
–
1
of
1
results
1
Sigma Aldrich 2-Oxo-2,3-dihydro-1H-benzoimidazole-5-boronic acid, pinacol ester
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.