accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONHOF

CAS RN 71083-06-2

IUPAC Name: ethyl 8-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate
Synonyms: ethyl 8-fluoro-4-oxo-1H-quinoline-3-carboxylate
Molecular Weight (g/mol): 235.21
Molecular Formula: C12H10FNO3
InChi Key: MPUYCZQHTGRPNE-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CNC2=C(F)C=CC=C2C1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results