accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 711-38-6

H3COOFFF

CAS RN 711-38-6

IUPAC Name: 2,2,2-trifluoro-1-(4-methoxyphenyl)ethan-1-one
Synonyms: 2,2,2-trifluoro-1-(4-methoxyphenyl)ethanone
Molecular Weight (g/mol): 204.15
Molecular Formula: C9H7F3O2
InChi Key: NCJZVRPXSSYDBG-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(=O)C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results