accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHO

CAS RN 71101-89-8

IUPAC Name: 4-butylcyclohexane-1-carboxylic acid
Synonyms: 4-butylcyclohexane-1-carboxylic acid
Molecular Weight (g/mol): 184.28
Molecular Formula: C11H20O2
InChi Key: BALGERHMIXFENA-UHFFFAOYSA-N
SMILES: CCCCC1CCC(CC1)C(O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results