CAS RN 71112-64-6
IUPAC Name:
bromo(phenanthren-9-yl)magnesium
Synonyms:
bromo(phenanthren-9-yl)magnesium
Molecular Weight (g/mol):
281.44
Molecular Formula:
C14H9BrMg
InChi Key:
ACMUZKQPXSHTGB-UHFFFAOYSA-M
SMILES:
Br[Mg]C1=CC2=C(C=CC=C2)C2=C1C=CC=C2
1
–
1
of
1
results
1
Sigma Aldrich N-Benzoyladenosine
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.