CAS RN 71133-14-7
IUPAC Name:
2-bromo-2,2-dichloroacetic acid
Synonyms:
bromodichloroacetic acid
Molecular Weight (g/mol):
207.83
Molecular Formula:
C2HBrCl2O2
InChi Key:
XSWVFEQKZFUULO-UHFFFAOYSA-N
SMILES:
OC(=O)C(Cl)(Cl)Br
1
–
1
of
1
results
1
Bromodichloroacetic Acid, SPEX CertiPrep™
CAS: 71133-14-7 Molecular Formula: C2HBrCl2O2 Molecular Weight (g/mol): 207.832 InChI Key: XSWVFEQKZFUULO-UHFFFAOYSA-N PubChem CID: 114809 IUPAC Name: 2-bromo-2,2-dichloroacetic acid SMILES: C(=O)(C(Cl)(Cl)Br)O