accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3

CAS RN 7116-95-2

IUPAC Name: 4-(propan-2-yl)-1,1'-biphenyl
Synonyms: 4-isopropylbiphenyl
Molecular Weight (g/mol): 196.29
Molecular Formula: C15H16
InChi Key: KWSHGRJUSUJPQD-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=C(C=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results