accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C

CAS RN 7116-97-4

IUPAC Name: 4-octyl-1,1'-biphenyl
Synonyms: 4-octyl-1,1'-biphenyl
Molecular Weight (g/mol): 266.43
Molecular Formula: C20H26
InChi Key: KCLBLDDJJJNGBC-UHFFFAOYSA-N
SMILES: CCCCCCCCC1=CC=C(C=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results