accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7118-66-3

IUPAC Name: 6-methyl-2-(4-methylphenyl)-1H-1,3-benzodiazole
Synonyms: 5-methyl-2-(4-methylphenyl)-3H-1,3-benzodiazole
Molecular Weight (g/mol): 222.29
Molecular Formula: C15H14N2
InChi Key: OPBSAVHQPYKEQP-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C1=NC2=CC=C(C)C=C2N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results