accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONONN

CAS RN 7119-95-1

IUPAC Name: 1-nitro-1H-pyrazole
Synonyms: 1-nitropyrazole
Molecular Weight (g/mol): 113.08
Molecular Formula: C3H3N3O2
InChi Key: TYNVOQYGXDUHRX-UHFFFAOYSA-N
SMILES: [O-][N+](=O)N1C=CC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results