accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 71229-85-1

IUPAC Name: 4-bromo-1-ethyl-1H-pyrazole
Synonyms: 4-bromo-1-ethylpyrazole
Molecular Weight (g/mol): 175.03
Molecular Formula: C5H7BrN2
InChi Key: IPMSARLBJARXSC-UHFFFAOYSA-N
SMILES: CCN1C=C(Br)C=N1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results