accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 71258-18-9

IUPAC Name: 1-benzyl-4-phenylpiperidine-4-carbonitrile hydrochloridyl
Synonyms: 1-benzyl-4-phenylpiperidine-4-carbonitrile hydrochloridyl
Molecular Weight (g/mol): 311.83
Molecular Formula: C19H20ClN2
InChi Key: NHLSXCISFQTVSW-UHFFFAOYSA-N
SMILES: [Cl].N#CC1(CCN(CC2=CC=CC=C2)CC1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results