accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONHO

CAS RN 7126-57-0

IUPAC Name: ethyl 4-formyl-1H-pyrrole-2-carboxylate
Synonyms: ethyl 4-formyl-1H-pyrrole-2-carboxylate
Molecular Weight (g/mol): 167.16
Molecular Formula: C8H9NO3
InChi Key: WVSAWXIWWNJTAV-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC(C=O)=CN1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results