accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 713125-22-5

IUPAC Name: 4-bromo-9,9-diphenyl-9H-fluorene
Synonyms: 4-bromo-9,9-diphenylfluorene
Molecular Weight (g/mol): 397.32
Molecular Formula: C25H17Br
InChi Key: PBWATBVKPGTOTB-UHFFFAOYSA-N
SMILES: BrC1=CC=CC2=C1C1=CC=CC=C1C2(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results